  • Search Results
  • Comparison of an Enzyme-Linked Immunosorbent Assay with an Immunochromatographic Assay for Detection of Lincomycin in Milk and Honey.
Comparison of an Enzyme-Linked Immunosorbent Assay with an Immunochromatographic Assay for Detection of Lincomycin in Milk and Honey.
Immunological investigations (2015-06-25)
Shanshan Cao, Shanshan Song, Liqiang Liu, Na Kong, Hua Kuang, Chuanlai Xu
ABSTRACT
An enzyme-linked immunosorbent assay (ELISA) and an immunochromatographic assay were constructed for the detection of lincomycin (LIN) in both milk and honey samples based on the monoclonal antibody named 5F6. The half-maximum inhibition of ELISA was 0.3 ng/mL after optimizing pH and ionic strength conditions; the limit of detection was 0.07 ng/mL. The cross-reactivity with clindamycin was 0.6%. LIN recovery in spiked milk and honey samples ranged from 84.6% to 115.6% with intra-assay coefficient variations of 1.7-25.4% and inter-assay coefficient variations of 2.7-8.9%. The detection limits were estimated as 2.1 µg/L for milk and 2.1 µg/kg for honey samples. The immunochromatographic assay revealed a LIN cut-off value of 10 ng/mL in PBS, 5 ng/mL in milk, and 120 ng/g in honey, and a visual lower detection limit of 2.5 ng/mL, 1 ng/mL and 30 ng/g in PBS, milk and honey, respectively. The immunochromatographic assay is preferred for large-scale practical application for its simpler pretreatment and satisfied sensitivity compared with ELISA assay.
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Sigma-Aldrich
1,2-Dichloroethane, anhydrous, 99.8%
Sigma-Aldrich
Kanamycin sulfate from Streptomyces kanamyceticus, powder, BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Streptomycin sulfate salt, powder
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥99%
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Succinic anhydride, ≥99% (GC)
Sigma-Aldrich
Kanamycin sulfate from Streptomyces kanamyceticus, meets USP testing specifications, powder
Sigma-Aldrich
Kanamycin sulfate from Streptomyces kanamyceticus, Animal Component-free
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98% (TLC)
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98.0% (NT)
Sigma-Aldrich
Erythromycin, potency: ≥850 μg per mg
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, tablet, 1 mg substrate per tablet
Sigma-Aldrich
Lincomycin hydrochloride, ≥95.0% (TLC)
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Lincomycin hydrochloride, ≥90% (TLC)
Sigma-Aldrich
Erythromycin, meets USP testing specifications
Sigma-Aldrich
Kanamycin sulfate Streptomyces kanamyceticus, powder, γ-irradiated
Sigma-Aldrich
Erythromycin standard solution, 1 mg/mL in H2O
Sigma-Aldrich
Lincomycin hydrochloride, BioReagent, suitable for cell culture
Sigma-Aldrich
Clindamycin 2-phosphate, aminoglycoside antibiotic