  • Search Results
  • Sequential solvent induced phase transition extraction for profiling of endogenous phytohormones in plants by liquid chromatography-mass spectrometry.
Sequential solvent induced phase transition extraction for profiling of endogenous phytohormones in plants by liquid chromatography-mass spectrometry.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-10-07)
Bao-Dong Cai, Er-Cui Ye, Bi-Feng Yuan, Yu-Qi Feng
ABSTRACT
In the current study, a novel method for high-throughput and sensitive determination of 12 phytohormones in plants was developed by using sequential solvent induced phase transition extraction (SIPTE) coupled with ultra-performance liquid chromatography-tandem mass spectrometry (UPLC-MS/MS). In sequential SIPTE, 0.1% formic acid (v/v) and 50mM NaHCO3 aqueous solution were used for enrichment and purification of alkaline and acidic phytohormones from the acetonitrile extract of plant tissues in sequence, in which hydrophobic solvent (toluene) was added to the acetonitrile aqueous mixture for driving the phase separation. Under optimized sequential SIPTE conditions, the phytohormones in acetonitrile extract of plant tissues could be effectively enriched and purified, which was in favor of the following UPLC-MS/MS analysis with less matrix effect. The phytohormones could be detected using the developed sequential SIPTE-UPLC-MS/MS method with the limits of the detection (LODs) ranging from 0.56 to 438.60pgmL(-1) and linear range over 2 orders of magnitude with correlation coefficients (r)>0.9970. The relative recoveries of the detected phytohormones were in the range of 85.1-114.6%. Finally, the proposed method was applied to simultaneous determination of endogenous phytohormones in different tissues of model plants (Oryza sativa and Arabidopsis thaliana) with small amount of sample size (5mg, fresh weight). The proposed method may be suitable for studying the distribution of phytohormones in model plants.
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Dichloromethane, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
Cyclohexane, anhydrous, 99.5%
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Dichloromethane, puriss. p.a., ACS reagent, reag. ISO, ≥99.9% (GC)
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Hexane, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)
Sigma-Aldrich
Cyclohexane, ACS reagent, ≥99%
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Ethyl acetate, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99.5% (GC)
Sigma-Aldrich
1-Octanol, anhydrous, ≥99%
Sigma-Aldrich
Butyl acetate, anhydrous, ≥99%
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Butyl acetate, ACS reagent, ≥99.5%