  • Search Results
  • Imaging mass spectrometry increased resolution using 2-mercaptobenzothiazole and 2,5-diaminonaphtalene matrices: application to lipid distribution in human colon.
Imaging mass spectrometry increased resolution using 2-mercaptobenzothiazole and 2,5-diaminonaphtalene matrices: application to lipid distribution in human colon.
Analytical and bioanalytical chemistry (2015-04-24)
Jone Garate, Roberto Fernández, Sergio Lage, Joan Bestard-Escalas, Daniel H Lopez, Rebeca Reigada, Sam Khorrami, Daniel Ginard, José Reyes, Isabel Amengual, Gwendolyn Barceló-Coblijn, José A Fernández
ABSTRACT
Imaging mass spectrometry is becoming a reference technique in the field of lipidomics, due to its ability to map the distribution of hundreds of species in a single run, along a tissue section. The next frontier is now achieving increasing resolution powers to offer cellular (or even sub-cellular) resolution. Thus, the new spectrometers are equipped with sophisticated optical systems to decrease the laser spot to <30 μm. Here, we demonstrate that by using the correct matrix (i.e., a matrix that maximizes ion detection and forms small crystals) and a careful preparation, it is possible to achieve resolutions of ∼5-10 μm, even with spectrometers equipped with non-optimal optics, which produces laser spots of 50 μm or even larger. As a proof of concept, we present images of distributions of lipids, both in positive and negative ion mode, over human colon endoscopic sections, recorded using 2-mercaptobenzothiazole for positive ion mode and 2,5-diaminonaphtalene for negative ion mode and an LTQ-Orbitrap XL, equipped with a matrix-assisted laser desorption ionization (MALDI) source that produces astigmatic laser spots. Graphical Abstract Imaging mass spectrometry is becoming an invaluable technique to complement traditional histology, but still higher resolutions are required. Here we deal with such issue.
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
2-Propanol, BioReagent, for molecular biology, ≥99.5%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Supelco
Hematoxylin
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
2-Mercaptobenzothiazole, 97%
Sigma-Aldrich
Hematoxylin, certified by the Biological Stain Commission
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
2,3-Diaminonaphthalene, ≥95% (HPLC), powder
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
2,3-Diaminonaphthalene, BioReagent, suitable for fluorescence, ≥98.0% (HPLC)
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%