  • Search Results
  • Evaluation of Mycelial Nutrients, Bioactive Compounds, and Antioxidants of Five Himalayan Entomopathogenic Ascomyceteous Fungi from India.
Evaluation of Mycelial Nutrients, Bioactive Compounds, and Antioxidants of Five Himalayan Entomopathogenic Ascomyceteous Fungi from India.
International journal of medicinal mushrooms (2015-11-13)
Sapan Kumar Sharma, Nandini Gautam, Narender Singh Atri
ABSTRACT
In this study, using standard methods, mycelial nutrients, bioactive compounds, and antioxidants were analyzed for the first time for five fungal species: Isaria sinclairii (Berk.) Lloyd, I. tenuipes Peck, I. japonica Yasuda, I. farinosa (Holmsk) Fr. and Cordyceps tuberculata (Lebert) Maire. All of these species were low in fat content and rich in protein, fiber, ash, and carbohydrates. Mineral elements (Fe, Mg, Cu, Mn, and Ca) were detected in appreciable amounts. All three types of fatty acids (saturated, monounsaturated, and polyunsaturated) as well as bioactive compounds (ascorbic acid, β-carotene, lycopene, phenolic compounds, and polysaccharides) were detected for each species. The investigated species showed high ferric-reducing antioxidant power as well as 2,2-diphenyl-1-picryl-hydrazyl radical scavenging activity. Although differences were observed in the values of each species, each species showed richness in one or more components.
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
Hexane, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Acetone, natural, ≥97%
Supelco
Acetone solution, certified reference material, 2000 μg/mL in methanol: water (9:1)
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Supelco
Acetone solution, contains 20.0 % (v/v) acetonitrile, suitable for HPLC
Supelco
Hexane, suitable for HPLC, ≥95%