  • Search Results
  • N-trimethyl chitosan (TMC)-modified microemulsions for improved oral bioavailability of puerarin: preparation and evaluation.
N-trimethyl chitosan (TMC)-modified microemulsions for improved oral bioavailability of puerarin: preparation and evaluation.
Drug delivery (2014-01-29)
Dehua Liao, Xinyi Liu, Wei Dai, Tiantian Tang, Ge Ou, Kai Zhang, Meng Han, Rongrong Kang, Sufang Yang, Daxiong Xiang
ABSTRACT
The aim of this research was to increase the oral bioavailability of puerarin by N-trimethyl chitosan-modified microemulsions (TMC-MEs) loaded with puerarin. Different concentrations of TMC-modified microemulsions were prepared in our study, and then evaluated for particle size, zeta potential, morphological observation and changes of the microenvironment polarity of inner oil core. It was shown that the zeta potential of the microemulsion was increased with the increasing concentration of TMC, and the peak value was achieved when the concentration of TMC was 3.0 mg/mL. The enhancement of the ratio of I(1)/I(3) (the ratio between the first band and the third band of the emission fluorescence spectrum of pyrene, I(1) = 373 nm, I(3) = 384 nm) indicated that polarity of the inner core of TMC-MEs was increased with the addition of the modifier. Pharmacokinetic studies demonstrated that after oral administration of puerarin N-trimethyl chitosan (TMC)-modified microemulsions (PUE-TMEs) and puerarin microemulsions (PUE-MEs) to rats at a dose of 100 mg/kg, relative bioavailability was enhanced about 6.8- and 1.2-fold, respectively, compared to puerarin suspension (PUE-SUS) as control. It indicated that the TMC-MEs could be used as an effective formulation for enhancing the oral bioavailability of puerarin.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, anhydrous, denatured
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
1-Methyl-2-pyrrolidinone, anhydrous, 99.5%
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Sigma-Aldrich
Propylene Glycol, meets USP testing specifications
Sigma-Aldrich
Iodomethane, purum, ≥99.0% (GC)
Sigma-Aldrich
Cycloheximide solution, Ready-Made Solution, microbial, 100 mg/mL in DMSO, 0.2 μm filtered
Sigma-Aldrich
Cycloheximide, ≥95% (HPLC)
Sigma-Aldrich
Iodomethane, contains copper as stabilizer, ReagentPlus®, 99%
Sigma-Aldrich
Propylene glycol, FCC, FG
Sigma-Aldrich
1,2-Propanediol, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Sodium iodide, 99.999% trace metals basis
Sigma-Aldrich
Iodomethane, contains copper as stabilizer, ReagentPlus®, 99.5%
Sigma-Aldrich
1,2-Propanediol, ReagentPlus®, 99%
Sigma-Aldrich
Isopropyl myristate, 98%
Sigma-Aldrich
1,2-Propanediol, puriss. p.a., ACS reagent, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Isopropyl myristate, ≥98%
Sigma-Aldrich
Cycloheximide, Biotechnology Performance Certified
Sigma-Aldrich
Sodium iodide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%