  • Search Results
  • The juxtamembrane sequence of the Hepatitis C virus polymerase can affect RNA synthesis and inhibition by allosteric polymerase inhibitors.
The juxtamembrane sequence of the Hepatitis C virus polymerase can affect RNA synthesis and inhibition by allosteric polymerase inhibitors.
Virus genes (2015-04-22)
Y Wen, X Lin, B Fan, C T Ranjith-Kumar, C C Kao
ABSTRACT
The Hepatitis C virus (HCV) RNA-dependent RNA polymerase (RdRp), nonstructural protein 5B (NS5B), is anchored in the membrane through a C-terminal helix. A sequence of ca. 12 residues that connects the catalytically competent portion of the RdRp and the C-terminal helix, the juxtamembrane sequence (JMS), has a poorly defined role in RdRp function in a large part since it is translated from a cis-acting RNA element (CRE) that is essential for HCV replication. Using a HCV replicon that transposed a second copy of CRE to the 3' UTR of the HCV replicon, we demonstrate that amino acid substitutions in the JMS were detrimental for HCV replicon replication. Substitutions in the JMS also resulted in a defect in de novo-initiated RNAs synthesis in vitro and in a cell-based reporter assay. A nonnucleoside inhibitor of the NS5B that binds to the catalytic pocket was less potent in inhibiting NS5B in the presence of JMS mutations. The JMS mutants exhibit reduced stability in thermodenaturation assays, suggesting that the JMS helps confer a more stable conformation to NS5B that could impact RNA synthesis.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Boron, crystalline, 1 cm, 99.7% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Boron, ≥95% (boron), amorphous powder
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
Imidazole, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Imidazole, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Imidazole, ACS reagent, ≥99% (titration)