  • Search Results
  • Isolation, Culture and Identification of Porcine Skeletal Muscle Satellite Cells.
Isolation, Culture and Identification of Porcine Skeletal Muscle Satellite Cells.
Asian-Australasian journal of animal sciences (2015-06-25)
Bo-Jiang Li, Ping-Hua Li, Rui-Hua Huang, Wen-Xing Sun, Han Wang, Qi-Fa Li, Jie Chen, Wang-Jun Wu, Hong-Lin Liu
ABSTRACT
The objective of this study was to establish the optimum protocol for the isolation and culture of porcine muscle satellite cells. Mononuclear muscle satellite cells are a kind of adult stem cell, which is located between the basal lamina and sarcolemma of muscle fibers and is the primary source of myogenic precursor cells in postnatal muscle. Muscle satellite cells are a useful model to investigate the mechanisms of muscle growth and development. Although the isolation and culture protocols of muscle satellite cells in some species (e.g. mouse) have been established successfully, the culture system for porcine muscle satellite cells is very limited. In this study, we optimized the isolation procedure of porcine muscle satellite cells and elaborated the isolation and culture process in detail. Furthermore, we characterized the porcine muscle satellite cells using the immunofluorecence. Our study provides a reference for the isolation of porcine muscle satellite cells and will be useful for studying the molecular mechanisms in these cells.
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Pricing and availability is not currently available.
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Pricing and availability is not currently available.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Pricing and availability is not currently available.
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Pricing and availability is not currently available.
SAFC
HEPES, PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Pricing and availability is not currently available.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Protease from Streptomyces griseus, powder, BioReagent, suitable for mouse embryo cell culture, ≥3.5 units/mg solid
Pricing and availability is not currently available.
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Pricing and availability is not currently available.
Sigma-Aldrich
Ethanesulfonic acid, 95%
Pricing and availability is not currently available.
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Pricing and availability is not currently available.
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Pricing and availability is not currently available.
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Ethanesulfonic acid solution, 70 wt. % in H2O
Pricing and availability is not currently available.
SAFC
HEPES, PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production, suitable for cell culture
Pricing and availability is not currently available.
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.