Identification of monofloral honeys using HPLC-ECD and chemometrics.
Food chemistry (2015-10-17)
Jing Zhao, Xiaojing Du, Ni Cheng, Lanzhen Chen, Xiaofeng Xue, Jing Zhao, Liming Wu, Wei Cao
ABSTRACT
A total of 77 jujube, longan and chaste honey samples were collected from 18 different areas of China. Thirteen types of phenolic acids in the honey samples were analysed using high-performance liquid chromatography with electrochemical detection (HPLC-ECD). Moreover, HPLC-ECD fingerprints of the monofloral honey samples were established. From the analysis of the HPLC-ECD fingerprints, common chromatography peak information was obtained, and principal component analysis and discriminant analysis were performed using selected common chromatography peak areas as variables. By comparing with phenolic acids as variables, using a chemometric analysis which is based on the use of common chromatography peaks as variables, 36 honey samples and 41 test samples could be correctly identified according to their floral origin.
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Gallic acid, 97.5-102.5% (titration)
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Caffeic acid, ≥98.0% (HPLC)
Sigma-Aldrich
p-Coumaric acid, ≥98.0% (HPLC)
Sigma-Aldrich
Chlorogenic acid, ≥95% (titration)
Sigma-Aldrich
trans-Ferulic acid, 99%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Rosmarinic acid, ≥98% (HPLC), from Rosemarinus officinalis L.
Sigma-Aldrich
4-Hydroxybenzoic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Ellagic acid, ≥95% (HPLC), powder, from tree bark
Sigma-Aldrich
4-Hydroxybenzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
3,4-Dihydroxybenzoic acid, ≥97.0% (T)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Sinapic acid, ≥98%, powder
Sigma-Aldrich
Syringic acid, ≥95% (HPLC)
Sigma-Aldrich
Vanillic acid, 97%
Sigma-Aldrich
Rosmarinic acid, 96%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
trans-Ferulic acid, ≥99%
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Vanillic acid, ≥97%, FG
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, NMR reference standard