  • Search Results
  • Sevoflurane inhibits the phosphorylation of ribosomal protein S6 in neonatal rat brain.
Sevoflurane inhibits the phosphorylation of ribosomal protein S6 in neonatal rat brain.
International journal of clinical and experimental medicine (2015-12-03)
Hong Tan, Chang-Lin Li, Lu Zhang, Zhou-Jing Yang, Xuan Zhao, Ying-Wei Wang
ABSTRACT
Besides neurotoxic effects, inhaled anesthetics might have other adverse effects on the developing brain. Ribosomal protein S6 (rpS6), the first identified ribosomal protein undergoing phosphorylation, has important physiological functions in regulating protein synthesis, cell proliferation, and glucose homeostasis. To date, the function of sevoflurane on rpS6 phosphorylation is unclear. In our present study, we showed that sevoflurane anesthesia inhibited rpS6 phosphorylation in cerebral cortex and CA1 region of the hippocampus. The activity of Akt was detected to be reduced within both cortical and hippocampal regions in the brain with the treatment of sevoflurane. However, the treatment seemed to have no effect on the phosphorylation of mammalian target of rapamycin (mTOR), a downstream effector of Akt. Sevoflurane had a paradoxical effect on ERK activity in the hippocampus and cerebral cortex. Last but not the least, Sevoflurane increased PP1 activity in the cerebral cortex and hippocampus. Thus, the exposure to sevoflurane inhibited dramatically the phosphorylation of rpS6 in neonatal rat brains. The inhibitory effect of sevoflurane on rpS6 phosphorylation might be mediated by the suppression on AKT activity at an mTOR-independent manner and the enhancement of PP1 activity.
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Sodium fluoride, ACS reagent, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%