  • Search Results
  • Proteomic Analysis of Isolated Plasma Membrane Fractions from the Mammary Gland in Lactating Cows.
Proteomic Analysis of Isolated Plasma Membrane Fractions from the Mammary Gland in Lactating Cows.
Journal of agricultural and food chemistry (2015-08-04)
Qiongxian Yan, Shaoxun Tang, Zhiliang Tan, Xuefeng Han, Chuanshe Zhou, Jinhe Kang, Min Wang
ABSTRACT
The mammary gland of dairy cows is a formidable lipid-synthesizing machine for lactation. This unique function depends on the activities of plasma membrane (PM) proteins in mammary cells. Little information is known about the expression profiles of PM proteins and their functions during the lactating process. This study investigated the proteome map of PM fractions of mammary gland in lactating cows using 1D-Gel-LC-MS/MS and identified 872 nonredundant proteins with 141 unknown proteins, wherein 215 were PM-associated proteins. Most of the PM-associated proteins were binding, transport, and catalytic proteins such as annexin proteins, heat shock proteins, integrins, RAS oncogene family members, and S100 calcium binding proteins. The PM-associated pathways such as caveolae-mediated endocytosis, leukocyte extravasation, aldosterone signaling in epithelial cells, and remodeling of epithelial adherens junctions were also significantly over-represented. Proteomic analysis revealed the characteristics and predicted functions of PM proteins isolated from the lactating bovine mammary gland. These results further provide experimental evidence for the presence of many proteins predicted in the annotated bovine genome. The data generated here also provide a reference for the PM-related functional research in the mammary gland of lactating cows.
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sucrose, ≥99.5%
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
SAFC
HEPES
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
SAFC
HEPES
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, Grade II, suitable for plant cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)