  • Search Results
  • Metabolic stress induces a Wnt-dependent cancer stem cell-like state transition.
Metabolic stress induces a Wnt-dependent cancer stem cell-like state transition.
Cell death & disease (2015-07-03)
E Lee, J Yang, M Ku, N H Kim, Y Park, C B Park, J-S Suh, E S Park, J I Yook, G B Mills, Y-M Huh, J-H Cheong
ABSTRACT
Reciprocal interactions between cancer cells and the tumor microenvironment drive multiple clinically significant behaviors including dormancy, invasion, and metastasis as well as therapy resistance. These microenvironment-dependent phenotypes share typical characteristics with cancer stem cells (CSC). However, it is poorly understood how metabolic stress in the confined tumor microenvironment contributes to the emergence and maintenance of CSC-like phenotypes. Here, we demonstrate that chronic metabolic stress (CMS) in a long-term nutrient deprivation induces a Wnt-dependent phenoconversion of non-stem cancer cells toward stem-like state and this is reflected in the transcriptome analysis. Addition of Wnt3a as well as transfection of dominant-negative Tcf4 establishes an obligatory role for the Wnt pathway in the acquisition of CSC-like characteristics in response to metabolic stress. Furthermore, systematic characterization for multiple single cell-derived clones and negative enrichment of CD44+/ESA+ stem-like cancer cells, all of which recapitulate stem-like cancer characteristics, suggest stochastic adaptation rather than selection of pre-existing subclones. Finally, CMS in the tumor microenvironment can drive a CSC-like phenoconversion of non-stem cancer cells through stochastic state transition dependent on the Wnt pathway. These findings contribute to an understanding of the metabolic stress-driven dynamic transition of non-stem cancer cells to a stem-like state in the tumor metabolic microenvironment.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Doxorubicin hydrochloride, 98.0-102.0% (HPLC)
Sigma-Aldrich
Propidium iodide solution, solution (1.0 mg/ml in water)
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Doxorubicin hydrochloride, suitable for fluorescence, 98.0-102.0% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Goat Anti-Mouse IgG Antibody, (H+L) FITC Conjugated, 2 mg/mL (after reconstitution), Chemicon®
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)