  • Search Results
  • A novel effect of DMOG on cell metabolism: direct inhibition of mitochondrial function precedes HIF target gene expression.
A novel effect of DMOG on cell metabolism: direct inhibition of mitochondrial function precedes HIF target gene expression.
Biochimica et biophysica acta (2015-07-06)
Alexander V Zhdanov, Irina A Okkelman, Fergus W J Collins, Silvia Melgar, Dmitri B Papkovsky
ABSTRACT
Abnormal accumulation of oncometabolite fumarate and succinate is associated with inhibition of mitochondrial function and carcinogenesis. By competing with α-ketoglutarate, oncometabolites also activate hypoxia inducible factors (HIFs), which makes oncometabolite mimetics broadly utilised in hypoxia research. We found that dimethyloxalylglycine (DMOG), a synthetic analogue of α-ketoglutarate, commonly used to induce HIF signalling, inhibits O2 consumption in cancer cell lines HCT116 and PC12, well before activation of HIF pathways. A rapid suppression of cellular respiration was accompanied by a decrease in histone H4 lysine 16 acetylation and not abolished by double knockdown of HIF-1α and HIF-2α. In agreement with this, production of NADH and state 3 respiration in isolated mitochondria were down-regulated by the de-esterified DMOG derivative, N-oxalylglycine. Exploring the roles of DMOG as a putative inhibitor of glutamine/α-ketoglutarate metabolic axis, we found that the observed suppression of OxPhos and compensatory activation of glycolytic ATP flux make cancer cells vulnerable to combined treatment with DMOG and inhibitors of glycolysis. On the other hand, DMOG treatment impairs deep cell deoxygenation in 3D tissue culture models, demonstrating a potential to relieve functional stress imposed by deep hypoxia on tumour, ischemic or inflamed tissues. Indeed, using a murine model of colitis, we found that O2 availability in the inflamed colon tissue rapidly increased after application of DMOG, which could contribute to the known therapeutic effect of this compound. Overall, our results provide new insights into the relationship between mitochondrial function, O2 availability, metabolic reprogramming and associated diseases.
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
D-Mannitol, ≥98%
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, for molecular biology, ≥97.0%
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Sodium butyrate, ≥98.5% (GC)
Sigma-Aldrich
Protease Inhibitor Cocktail powder, for general use, lyophilized powder
Sigma-Aldrich
Sucrose, ≥99.5%
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Adenine, ≥99%
Sigma-Aldrich
Acrylamide, for molecular biology, ≥99% (HPLC)
Sigma-Aldrich
Sodium butyrate, 98%
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
Sucrose, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
SAFC
HEPES
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99% (HPLC), powder
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered
Sigma-Aldrich
Adenine, BioReagent, suitable for cell culture
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
DMOG, ≥98% (HPLC)