  • Search Results
  • Solar-Energy Driven Simultaneous Saccharification and Fermentation of Starch to Bioethanol for Fuel-Cell Applications.
Solar-Energy Driven Simultaneous Saccharification and Fermentation of Starch to Bioethanol for Fuel-Cell Applications.
ChemSusChem (2015-09-10)
Betina Tabah, Indra Neel Pulidindi, Venkateswara Rao Chitturi, Leela Mohana Reddy Arava, Aharon Gedanken
ABSTRACT
A solar reactor was designed to perform the conversion of starch to ethanol in a single step. An aqueous starch solution (5 wt %) was fed into the reactor bed charged with Baker's yeast (Saccharomyces cerevisiae) and amylase, resulting in approximately 2.5 wt % ethanol collected daily (ca. 25 mL day(-1) ). A significant amount of ethanol (38 g) was collected over 63 days, corresponding to 84 % of the theoretical yield. The production of ethanol without additional energy input highlights the significance of this new process. The ethanol produced was also demonstrated as a potential fuel for direct ethanol fuel cells. Additionally, the secondary metabolite glycerol was fully reduced to a value-added product 1,3-propanediol, which is the first example of a fungal strain (Baker's yeast) converting glycerol in situ to 1,3-propanediol.
Sigma-Aldrich
2-Propanol, BioReagent, for molecular biology, ≥99.5%
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.05 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
Deuterium oxide, "100%", ≥99.96 atom % D
Sigma-Aldrich
Deuterium oxide, 99 atom % D
Sigma-Aldrich
2-Propanol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Deuterium oxide, filtered, 99.8 atom % D
Sigma-Aldrich
Deuterium oxide, 99.8 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 1 % (w/w) 3-(trimethylsilyl)-1-propanesulfonic acid, sodium salt (DSS)
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.75 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Deuterium oxide, 70 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, glass distilled
Sigma-Aldrich
Deuterium oxide, "100%", 99.990 atom % D
Sigma-Aldrich
Deuterium oxide, standard, 99.98 atom %±0.01 atom % D
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
Deuterium oxide, extra, 99.994 atom % D
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Deuterium oxide, 60 atom % D
Sigma-Aldrich
2-Propanol, 99.5%, HPLC grade