  • Search Results
  • Isotope-labeled differential profiling of metabolites using N-benzoyloxysuccinimide derivatization coupled to liquid chromatography/high-resolution tandem mass spectrometry.
Isotope-labeled differential profiling of metabolites using N-benzoyloxysuccinimide derivatization coupled to liquid chromatography/high-resolution tandem mass spectrometry.
Rapid communications in mass spectrometry : RCM (2015-10-16)
Michel Wagner, Leanne B Ohlund, Tze Chieh Shiao, Amélie Vézina, Borhane Annabi, René Roy, Lekha Sleno
ABSTRACT
An isotopic labeling strategy based on derivatizing amine-containing metabolites has been developed using light ((12) C6 ) and heavy ((13) C6 ) N-benzoyloxysuccinimide reagents for semi-targeted metabolomic applications. Differentially labeled samples were combined and analyzed simultaneously by liquid chromatography/high-resolution tandem mass spectrometry (LC/HR-MS/MS) to compare relative amounts of amine-containing metabolites. The selectivity of the reaction was determined with model metabolites and was shown to also be applicable to thiol and phenol moieties. The potential for relative quantitation was evaluated in cell extracts and the method was then applied to quantify metabolic perturbations occurring in human cultured cells under normal vs. oxidative stress conditions. A total of 279 derivatized features were detected in HL60 cell extracts, 77 of which yielded significant concentration changes upon oxidative stress treatment. Based on accurate mass measurements and MS/MS spectral matching with reference standard solutions, 10 metabolites were clearly identified. Derivatized compounds were found to have diagnostic fragment ions from the reagent itself, as well as structurally informative ions useful for metabolite identification. This simple derivatization reaction can be applied to the relative quantitation of amine-, thiol- and phenol-containing compounds, with improved sensitivity and chromatographic peak shapes due to the increased hydrophobicity of polar metabolites not readily amenable to reversed-phase LC/MS analysis.
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 0.1 % (v/v) formic acid, 5 % (v/v) water, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 40.0% 2-propanol, 0.05% formic acid
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
DL-Histidine, ≥99% (TLC)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
DL-Aspartic acid, ≥99% (TLC)
Sigma-Aldrich
L-Glutathione oxidized disodium salt, suitable for cell culture, BioReagent
Sigma-Aldrich
L-Glutathione oxidized disodium salt, ≥98%, powder
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
DL-Tryptophan, ≥99% (HPLC)
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
DL-Tryptophan, ≥99% (HPLC)
Sigma-Aldrich
Trypan Blue, Dye content 60 %
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
SAFC
L-Threonine
Sigma-Aldrich
L-Threonine, BioXtra, ≥99.5% (NT)
SAFC
Glycine
Sigma-Aldrich
2-Aminoisobutyric acid, 98%
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
L-Threonine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Threonine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC