  • Search Results
  • Characterization of dehydroascorbate-mediated modification of glutaredoxin by mass spectrometry.
Characterization of dehydroascorbate-mediated modification of glutaredoxin by mass spectrometry.
Journal of mass spectrometry : JMS (2015-12-05)
Aurore Flandrin, Sebastien Allouche, Yoann Rolland, François-Olivier McDuff, J Richard Wagner, Klaus Klarskov
ABSTRACT
Ascorbate is as a potent antioxidant in vivo protecting the organism against oxidative stress. In this process, ascorbate is oxidized in two steps to dehydroascorbate (DHA), which if not efficiently reduced back to ascorbate decomposes irreversibly to a complex mixture of products. We demonstrate that a component of this mixture specifically reacts with the thiol group of cysteine residues at physiological pH to give a protein adduct involving the addition of a 5-carbon fragment of DHA (+112 Da). Incubations of glutaredoxin-1 expressed in Escherichia coli and dehydroascorbate revealed abundant adducts of +112, +224 and +336 Da due to the addition of one, two and three conjugation products of DHA, respectively. ESI-MS of carbamidomethylated glutaredoxin-1 before incubation with DHA, deuterium exchange together with tandem mass spectrometry analysis and LC-ESIMS/MS of modified peptides confirmed structure and sites of modification in the protein. Modification of protein thiols by a DHA-derived product can be involved in oxidative stress-mediated cellular toxicity.
Sigma-Aldrich
Iodoacetamide, ≥99% (NMR), crystalline
Sigma-Aldrich
Iodoacetamide, BioUltra
Sigma-Aldrich
Iodoacetamide, Single use vial of 56 mg
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.05 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D
Sigma-Aldrich
Deuterium oxide, "100%", 99.990 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 1 % (w/w) 3-(trimethylsilyl)-1-propanesulfonic acid, sodium salt (DSS)
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D
Sigma-Aldrich
Deuterium oxide, "100%", ≥99.96 atom % D
Sigma-Aldrich
Deuterium oxide, 99.8 atom % D
Sigma-Aldrich
Deuterium oxide, 99 atom % D
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Sigma-Aldrich
Deuterium oxide, extra, 99.994 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, glass distilled
Sigma-Aldrich
Deuterium oxide, standard, 99.98 atom %±0.01 atom % D
Sigma-Aldrich
Deuterium oxide, 70 atom % D
Sigma-Aldrich
Deuterium oxide, 60 atom % D
Sigma-Aldrich
Deuterium oxide, filtered, 99.8 atom % D
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
SAFC
Iodoacetamide
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.75 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%