  • Search Results
  • Identification and Characterization of the Androgen Receptor From the American Alligator, Alligator mississippiensis.
Identification and Characterization of the Androgen Receptor From the American Alligator, Alligator mississippiensis.
Endocrinology (2015-05-15)
Shinichi Miyagawa, Ryohei Yatsu, Satomi Kohno, Brenna M Doheny, Yukiko Ogino, Hiroshi Ishibashi, Yoshinao Katsu, Yasuhiko Ohta, Louis J Guillette, Taisen Iguchi
ABSTRACT
Androgens are essential for the development, reproduction, and health throughout the life span of vertebrates, particularly during the initiation and maintenance of male sexual characteristics. Androgen signaling is mediated by the androgen receptor (AR), a member of the steroid nuclear receptor superfamily. Mounting evidence suggests that environmental factors, such as exogenous hormones or contaminants that mimic hormones, can disrupt endocrine signaling and function. The American alligator (Alligator mississippiensis), a unique model for ecological research in that it exhibits environment-dependent sex determination, is oviparous and long lived. Alligators from a contaminated environment exhibit low reproductive success and morphological disorders of the testis and phallus in neonates and juveniles, both associated with androgen signaling; thus, the alterations are hypothesized to be related to disrupted androgen signaling. However, this line of research has been limited because of a lack of information on the alligator AR gene. Here, we isolated A mississippiensis AR homologs (AmAR) and evaluated receptor-hormone/chemical interactions using a transactivation assay. We showed that AmAR responded to all natural androgens and their effects were inhibited by cotreatment with antiandrogens, such as flutamide, p,p'-dichlorodiphenyldichloroethylene, and vinclozolin. Intriguingly, we found a spliced form of the AR from alligator cDNA, which lacks seven amino acids within the ligand-binding domain that shows no response to androgens. Finally, we have initial data on a possible dominant-negative function of the spliced form of the AR against androgen-induced AmAR.
Sigma-Aldrich
Phenol, contains hypophosphorous as stabilizer, loose crystals, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, for molecular biology
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Phenol solution, Saturated with 0.1 M citrate buffer, pH 4.3 ± 0.2, BioReagent, for molecular biology
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC), crystalline (detached)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Phenol, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Phenol, natural, 97%, FG
Sigma-Aldrich
Aldosterone, ≥95% (HPLC)