  • Search Results
  • Nigrosome 1 Detection at 3T MRI for the Diagnosis of Early-Stage Idiopathic Parkinson Disease: Assessment of Diagnostic Accuracy and Agreement on Imaging Asymmetry and Clinical Laterality.
Nigrosome 1 Detection at 3T MRI for the Diagnosis of Early-Stage Idiopathic Parkinson Disease: Assessment of Diagnostic Accuracy and Agreement on Imaging Asymmetry and Clinical Laterality.
AJNR. American journal of neuroradiology (2015-08-22)
Y Noh, Y H Sung, J Lee, E Y Kim
ABSTRACT
In the early stages of idiopathic Parkinson disease, motor symptoms are usually asymmetric. We aimed to assess the feasibility of nigrosome 1 detection at 3T MR imaging to analyze the agreement of its asymmetry and clinical laterality. High-resolution 3D multiecho imaging was performed at 3T MR imaging in 13 healthy subjects and 24 patients with idiopathic Parkinson disease confirmed by N-3-fluoropropyl-2-β-carbomethoxy-3-β-(4-iodophenyl) nortropane ((18)F-FP-CIT) PET. The nigrosome 1 detection findings by using the MR imaging data were rated as "normal," "possibly abnormal," and "abnormal" by 2 independent reviewers. The degree of (18)F-FP-CIT binding was visually assessed in the caudate nucleus and putamen on PET images. Clinical laterality was evaluated by scores of the Unified Parkinson Disease Rating Scale, Part III. Asymmetry of the affected nigrosome 1 and the degree of (18)F-FP-CIT binding were analyzed for agreement with clinical laterality. The diagnostic sensitivity, specificity, and accuracy of the nigrosome 1 detection at 3T MR imaging was 100%, 84.6%, and 94.6%, respectively. Interrater agreements for the abnormality and asymmetry of nigrosome 1 were excellent (κ = 0.863 and 0.835, respectively). In patients with idiopathic Parkinson disease, the agreement of asymmetry between clinical laterality and nigrosome 1 detection was good (κ = 0.724). The degree of the (18)F-FP-CIT PET binding showed fair agreement (κ = 0.235) with clinical laterality. The abnormality involving nigrosome 1 can be detected at 3T MR imaging with an accuracy of 94.6%. The clinical laterality is in high concordance with the laterality of the nigrosome 1 detection at 3T (κ = 0.724).
Sigma-Aldrich
Tantalum(V) ethoxide, 99.98% trace metals basis
Tantalum(V) ethoxide, packaged for use in deposition systems