  • Search Results
  • The control of tomato fruit elongation orchestrated by sun, ovate and fs8.1 in a wild relative of tomato.
The control of tomato fruit elongation orchestrated by sun, ovate and fs8.1 in a wild relative of tomato.
Plant science : an international journal of experimental plant biology (2015-08-11)
Shan Wu, Josh P Clevenger, Liang Sun, Sofia Visa, Yuji Kamiya, Yusuke Jikumaru, Joshua Blakeslee, Esther van der Knaap
ABSTRACT
Within the cultivated tomato germplasm, sun, ovate and fs8.1 are the three predominant QTLs controlling fruit elongation. Although SUN and OVATE have been cloned, their role in plant growth and development are not well understood. To compare and contrast the effects of the three QTLs in a homogeneous background, we developed near isogenic lines (NILs) in the wild species Solanum pimpinellifolium LA1589 background. We carried out detailed morphological characterization of reproductive and vegetative organs in the single, double and triple NILs and determined the epistatic interactions of the three loci affecting fruit shape. The phenotypic evaluations demonstrated that the three loci regulate unique aspects of ovary and fruit elongation and in different temporal manners. The strongest effect on organ shape was caused by sun. In addition to fruit shape, sun also affected leaf and sepal elongation and stem thickness. The synergistic interaction between sun and ovate or fs8.1 suggested that the pathways involving SUN, OVATE and the gene(s) underlying fs8.1 may converge at a common node. The results of an extensive profiling analysis suggested that the degree of fruit elongation was not related to the accumulation of any of the classical hormones.
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
2-Propanol, BioReagent, for molecular biology, ≥99.5%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Sodium phosphate, 96%
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Nitrogen, ≥99.998%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
3-Indolepropionic acid, ReagentPlus®, 99%
Sigma-Aldrich
Indole-3-propionic acid, ≥99.0% (T)
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Supelco
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Indole-3-propionic acid, ≥97.0% (T)
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Methanol solution, contains 0.1 % (v/v) trifluoroacetic acid, 5 % (v/v) water, suitable for HPLC
Sigma-Aldrich
Methanol solution, (Methanol:Dimethyl sulfoxide 1:1 (v/v))
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
2-Propanol, 99.5%, HPLC grade