  • Search Results
  • Simultaneous determination of carboprost methylate and its active metabolite carboprost in dog plasma by liquid chromatography-tandem mass spectrometry with positive/negative ion-switching electrospray ionization and its application to a pharmacokinetic study.
Simultaneous determination of carboprost methylate and its active metabolite carboprost in dog plasma by liquid chromatography-tandem mass spectrometry with positive/negative ion-switching electrospray ionization and its application to a pharmacokinetic study.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-07-08)
Lei Yin, Xiangjun Meng, Xiaotong Zhou, Tinglan Zhang, Heping Sun, Zhichao Yang, Bo Yang, Ning Xiao, J Paul Fawcett, Yan Yang, Jingkai Gu
ABSTRACT
A liquid chromatography-tandem mass spectrometric (LC-MS/MS) method using positive/negative electrospray ionization (ESI) switching for the simultaneous quantitation of carboprost methylate and carboprost in dog plasma has been developed and validated. After screening, the esterase inhibitor, dichlorvos was added to the whole blood at a ratio of 1:99 (v/v) to stabilize carboprost methylate during blood collection, sample storage and LLE. Indomethacin was added to plasma to inhibit prostaglandins synthesis after sampling. After liquid-liquid extraction of 500μL plasma with ethyl ether-dichloromethane (75:25, v/v), analytes and internal standard (IS), alprostadil-d4, were chromatographed on a CAPCELL PAK Phenyl column (150×2.0mm, 5μm) using acetonitrile-5mM ammonium acetate as mobile phase. Carboprost methylate was detected by positive ion electrospray ionization followed by multiple reaction monitoring (MRM) of the transition at m/z 400.5→329.3; the carboprost and IS were detected by negative ion electrospray ionization followed by MRM of the transitions at m/z 367.2→323.2, and 357.1→321.2, respectively. The method was linear for both analytes in the concentration range 0.05-30ng/mL with intra- and inter-day precisions (as relative standard deviation) of ≤6.75% and accuracy (as relative error) of ≤7.21% and limit of detection (LOD) values were 10 and 20pg/mL, respectively. The method was successfully applied to a pharmacokinetic study of the analytes in beagle dogs after intravaginal administration of a suppository containing 0.5mg carboprost methylate.
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
Paraoxon-ethyl, ≥90%, oil
Sigma-Aldrich
2-Propanol, 99.5%, HPLC grade
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
2-Propanol, BioReagent, for molecular biology, ≥99.5%
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile
Sigma-Aldrich
Diisopropylfluorophosphate
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 0.1 % (v/v) formic acid, 5 % (v/v) water, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 40.0% 2-propanol, 0.05% formic acid