  • Search Results
  • Targeted metabolomics in the expanded newborn screening for inborn errors of metabolism.
Targeted metabolomics in the expanded newborn screening for inborn errors of metabolism.
Molecular bioSystems (2015-02-18)
Emanuela Scolamiero, Carla Cozzolino, Lucia Albano, Antonella Ansalone, Marianna Caterino, Graziella Corbo, Maria Grazia di Girolamo, Cristina Di Stefano, Adriano Durante, Giovanni Franzese, Ignazio Franzese, Giovanna Gallo, Paolo Giliberti, Laura Ingenito, Giovanni Ippolito, Basilio Malamisura, Pietro Mazzeo, Antonella Norma, Daniela Ombrone, Giancarlo Parenti, Silvana Pellecchia, Rita Pecce, Ippolito Pierucci, Roberta Romanelli, Anna Rossi, Massimo Siano, Teodoro Stoduto, Guglielmo R D Villani, Generoso Andria, Francesco Salvatore, Giulia Frisso, Margherita Ruoppolo
ABSTRACT
Inborn errors of metabolism are genetic disorders due to impaired activity of enzymes, transporters, or cofactors resulting in accumulation of abnormal metabolites proximal to the metabolic block, lack of essential products or accumulation of by-products. Many of these disorders have serious clinical consequences for affected neonates, and an early diagnosis allows presymptomatic treatment which can prevent severe permanent sequelae and in some cases death. Expanded newborn screening for these diseases is a promising field of targeted metabolomics. Here we report the application, between 2007 and 2014, of this approach to the identification of newborns in southern Italy at risk of developing a potentially fatal disease. The analysis of amino acids and acylcarnitines in dried blood spots by tandem mass spectrometry revealed 24 affected newborns among 45,466 infants evaluated between 48 and 72 hours of life (overall incidence: 1 : 1894). Diagnoses of newborns with elevated metabolites were confirmed by gas chromatography-mass spectrometry, biochemical studies, and genetic analysis. Five infants were diagnosed with medium-chain acyl CoA dehydrogenase deficiency, 1 with methylmalonic acidemia with homocystinuria type CblC, 2 with isolated methylmalonic acidemia, 1 with propionic acidemia, 1 with isovaleric academia, 1 with isobutyryl-CoA dehydrogenase deficiency, 1 with beta ketothiolase deficiency, 1 with short branched chain amino acid deficiency, 1 with 3-methlycrotonyl-CoA carboxylase deficiency, 1 with formimino-transferase cyclodeaminase deficiency, and 1 with cystathionine-beta-synthase deficiency. Seven cases of maternal vitamin B12 deficiency and 1 case of maternal carnitine uptake deficiency were detected. This study supports the widespread application of metabolomic-based newborn screening for these genetic diseases.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, anhydrous, denatured
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
1-Butanol, anhydrous, 99.8%
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Supelco
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, with 1% trimethylchlorosilane, for GC derivatization, LiChropur
Sigma-Aldrich
1-Butanol, for molecular biology, ≥99%
Sigma-Aldrich
N,O-Bis(trimethylsilyl)trifluoroacetamide, ≥99%
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Butyl alcohol, natural, ≥99.5%, FCC, FG
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydroxylamine hydrochloride, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Tropic acid, 98%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG