  • Search Results
  • A new target region for changing the substrate specificity of amine transaminases.
A new target region for changing the substrate specificity of amine transaminases.
Scientific reports (2015-06-02)
Li-Jun Guan, Jun Ohtsuka, Masahiko Okai, Takuya Miyakawa, Tomoko Mase, Yuehua Zhi, Feng Hou, Noriyuki Ito, Akira Iwasaki, Yoshihiko Yasohara, Masaru Tanokura
ABSTRACT
(R)-stereospecific amine transaminases (R-ATAs) are important biocatalysts for the production of (R)-amine compounds in a strict stereospecific manner. An improved R-ATA, ATA-117-Rd11, was successfully engineered for the manufacture of sitagliptin, a widely used therapeutic agent for type-2 diabetes. The effects of the individual mutations, however, have not yet been demonstrated due to the lack of experimentally determined structural information. Here we describe three crystal structures of the first isolated R-ATA, its G136F mutant and engineered ATA-117-Rd11, which indicated that the mutation introduced into the 136(th) residue altered the conformation of a loop next to the active site, resulting in a substrate-binding site with drastically modified volume, shape, and surface properties, to accommodate the large pro-sitagliptin ketone. Our findings provide a detailed explanation of the previously reported molecular engineering of ATA-117-Rd11 and propose that the loop near the active site is a new target for the rational design to change the substrate specificity of ATAs.
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Pyridoxal 5′-phosphate hydrate, ≥98%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Pyridoxal 5′-phosphate monohydrate, ≥97.0% (NT)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Pyridoxal 5′-phosphate hydrate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl