  • Search Results
  • The effect of losartan on differential reflex control of sympathetic nerve activity in chronic kidney disease.
The effect of losartan on differential reflex control of sympathetic nerve activity in chronic kidney disease.
Journal of hypertension (2015-03-13)
Yimin Yao, Cara M Hildreth, Melissa M Farnham, Manash Saha, Qi-Jian Sun, Paul M Pilowsky, Jacqueline K Phillips
ABSTRACT
The effect of angiotensin II type I receptor (AT1R) inhibition on the pattern of reflex sympathetic nerve activity (SNA) to multiple target organs in the Lewis polycystic kidney (LPK) rat model of chronic kidney disease was determined. Mean arterial pressure (MAP), splanchnic SNA (sSNA), renal SNA (rSNA) and lumbar SNA (lSNA) were recorded in urethane-anaesthetized LPK and Lewis controls (total n = 39). Baroreflex, peripheral and central chemoreflex, and somatosensory reflex control of SNA (evoked by phenylephrine/sodium nitroprusside infusion, 10% O2 in N2 or 100% N2 ventilation, 5% CO2 ventilation and sciatic nerve stimulation, respectively) were determined before and after administration of losartan (AT1R antagonist 3 mg/kg, intravenous). Baseline MAP was higher in LPK rats and baroreflex control of sSNA and rSNA, but not lSNA, was reduced. Losartan reduced MAP in both strains and selectively improved baroreflex gain for sSNA (-1.2 ± 0.1 vs. -0.7 ± 0.07 %/mmHg; P < 0.05) in LPK. The peripheral and central chemoreflex increased MAP and all SNA in Lewis controls, but reduced or had no effect on these parameters, respectively, in LPK. The SNA response to somatosensory stimulation was biphasic, with latency to second peak less in LPK. Losartan ameliorated the depressor and sympathoinhibitory responses to peripheral chemoreflex stimulation in the LPK, but did not alter the central chemoreflex or somatosympathetic responses. Inhibition of the AT1R selectively improved baroreflex control of sSNA and peripheral chemoreflex control of all three sympathetic nerve outflows in the LPK rat, suggesting these anomalies in reflex function are driven in part by angiotensin II.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Dibutyl phthalate, 99%
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Pancuronium bromide
Sigma-Aldrich
3-[(1R)-1-Hydroxy-2-(methylamino)ethyl]phenol, AldrichCPR
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Creatinine, anhydrous, ≥98%
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, BioXtra, ≥99.0% (T)
Sigma-Aldrich
DBP, 98% (HPLC)