  • Search Results
  • Purity Determination of Acetaldehyde in an Acetaldehyde Certified Reference Material.
Purity Determination of Acetaldehyde in an Acetaldehyde Certified Reference Material.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2015-06-13)
Taichi Yamazaki, Takuro Watanabe, Satoe Nakamura, Kenji Kato
ABSTRACT
Acetaldehyde is regulated as a toxic substance in various fields, and the method for monitoring or analysis of acetaldehyde is important. However, handling is difficult because of the high reactivity and low boiling point of acetaldehyde. Therefore, a reference material for high purity acetaldehyde with high accuracy was not available. Although the measuring method of acetaldehyde as a reagent is published in the Japanese Industrial Standard (JIS) where the specification of acetaldehyde purity is more than 80%, the analytical method described in JIS is not enough for an accuracy purity determination method. In this research, the high precision purity determination method for development of a certified reference material (CRM) of acetaldehyde was examined. By controlling the volatility and reactivity of acetaldehyde, we established the purity determination method of acetaldehyde with a relative standard uncertainty of less than 0.3%. Furthermore, this method was applied to develop a high purity acetaldehyde CRM with an expanded uncertainty of 0.005 kg kg(-1) (k = 2).
Sigma-Aldrich
1,4-Dioxane, anhydrous, 99.8%
Sigma-Aldrich
Acetaldehyde, ACS reagent, ≥99.5%
Sigma-Aldrich
Chlorobenzene, anhydrous, 99.8%
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetaldehyde, natural, FG
Sigma-Aldrich
Crotonaldehyde, predominantly trans, ≥99%, contains 0.1-0.2% BHT as stabilizer, 1% H2O as stabilizer
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in H2O
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Crotonaldehyde, mixture of cis and trans, ratio of cis- and trans-isomers (~1:20), ≥99.5% (GC)
Sigma-Aldrich
Acetaldehyde solution, natural, 50 wt. % ethanol, FG
Sigma-Aldrich
Acetaldehyde solution, 50 wt. % in ethanol
Sigma-Aldrich
Acetaldehyde, ≥99%, meets FCC analytical specification
Sigma-Aldrich
Acetaldehyde solution, 5 M in THF
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in isopropanol
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O