  • Search Results
  • Risk Stratification of Thyroid Incidentalomas Found on PET/CT: The Value of Iodine Content on Noncontrast Computed Tomography.
Risk Stratification of Thyroid Incidentalomas Found on PET/CT: The Value of Iodine Content on Noncontrast Computed Tomography.
Thyroid : official journal of the American Thyroid Association (2015-09-04)
Dongwoo Kim, Sang Hyun Hwang, Jongtae Cha, Kwanhyeong Jo, Narae Lee, Mijin Yun
ABSTRACT
The Hounsfield unit (HU) ratio of thyroid nodules was assessed compared to the contralateral thyroid lobe on noncontrast computed tomography (CT) to stratify further the risk of malignancy in thyroid incidentalomas found on 18F-fluorodeoxyglucose ((18)F-FDG) positron emission tomography/CT (PET/CT). This retrospective analysis included 82 patients who had thyroid incidentalomas on PET/CT in 2011. On PET/CT, the maximal standardized uptake value ratios of the thyroid nodule compared to liver (T/BSUV) and the HU ratios of the thyroid nodule compared to contralateral thyroid lobe (T/BHU) were calculated. Diagnostic performances of the T/BSUV and T/BHU were compared. The area under the curve of T/BHU was higher than that of T/BSUV (0.941 vs. 0.689, p < 0.0001). The sensitivity, specificity, and accuracy of T/BHU were significantly higher than those of T/BSUV (100% vs. 77.8%, p = 0.0313; 80.0% vs. 60.0%, p = 0.0433; 86.6% vs. 65.9%, p = 0.0041, respectively). The risk of malignancy was much higher (71.1%) in nodules with a T/BHU cutoff value ≤0.68, whereas it was 0% in nodules with a T/BHU of >0.68. In this study, there were 18 nodules with nondiagnostic (n = 7) or atypia of undetermined significance or follicular lesion of undetermined significance cytologies (n = 11) after fine-needle aspiration biopsy (FNAB). When the T/BHU cutoff value was applied, three (60%) of the five nodules with a T/BHU of ≤0.68 were found to be papillary carcinomas. The remaining 13 nodules with a T/BHU of >0.68 were all benign with a risk of malignancy of 0%. T/BHU is a simple and effective parameter to stratify the risk of malignancy in thyroid incidentalomas found on PET/CT. This may be of clinical relevance in those nodules with nondiagnostic or undetermined significance cytologies upon FNAB in the scheme of current clinical practice.
Sigma-Aldrich
Tantalum(V) ethoxide, 99.98% trace metals basis
Tantalum(V) ethoxide, packaged for use in deposition systems