  • Search Results
  • The a subunit of the A1AO ATP synthase of Methanosarcina mazei Gö1 contains two conserved arginine residues that are crucial for ATP synthesis.
The a subunit of the A1AO ATP synthase of Methanosarcina mazei Gö1 contains two conserved arginine residues that are crucial for ATP synthesis.
Biochimica et biophysica acta (2015-03-01)
Carolin Gloger, Anna-Katharina Born, Martin Antosch, Volker Müller
ABSTRACT
Like the evolutionary related F1FO ATP synthases and V1VO ATPases, the A1AO ATP synthases from archaea are multisubunit, membrane-bound transport machines that couple ion flow to the synthesis of ATP. Although the subunit composition is known for at least two species, nothing is known so far with respect to the function of individual subunits or amino acid residues. To pave the road for a functional analysis of A1AO ATP synthases, we have cloned the entire operon from Methanosarcina mazei into an expression vector and produced the enzyme in Escherichia coli. Inverted membrane vesicles of the recombinants catalyzed ATP synthesis driven by NADH oxidation as well as artificial driving forces. [Formula: see text] as well as ΔpH were used as driving forces which is consistent with the inhibition of NADH-driven ATP synthesis by protonophores. Exchange of the conserved glutamate in subunit c led to a complete loss of ATP synthesis, proving that this residue is essential for H+ translocation. Exchange of two conserved arginine residues in subunit a has different effects on ATP synthesis. The role of these residues in ion translocation is discussed.
Sigma-Aldrich
Chloramphenicol, ≥98% (HPLC)
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Sigma-Aldrich
Chloramphenicol, BioReagent, suitable for plant cell culture
Sigma-Aldrich
Ampicillin, meets USP testing specifications
Sigma-Aldrich
Chloramphenicol, meets USP testing specifications
Sigma-Aldrich
Chloramphenicol, γ-irradiated