  • Search Results
  • Cytochrome P450 2D6 and homeobox 13/interleukin-17B receptor: combining inherited and tumor gene markers for prediction of tamoxifen resistance.
Cytochrome P450 2D6 and homeobox 13/interleukin-17B receptor: combining inherited and tumor gene markers for prediction of tamoxifen resistance.
Clinical cancer research : an official journal of the American Association for Cancer Research (2008-09-17)
Matthew P Goetz, Vera J Suman, Fergus J Couch, Matthew M Ames, James M Rae, Mark G Erlander, Xiao-Jun Ma, Dennis C Sgroi, Carol A Reynolds, Wilma L Lingle, Richard M Weinshilboum, David A Flockhart, Zeruesenay Desta, Edith A Perez, James N Ingle
ABSTRACT
Genetic variation in cytochrome P450 2D6 (CYP2D6) and the gene expression ratio of the homeobox 13 (HOXB13) to interleukin-17B receptor (IL17BR) are associated with tamoxifen resistance. We sought to determine the combined effect of inherited (CYP2D6) and somatic (HOXB13/IL17BR) gene variation in tamoxifen-treated breast cancer. Retrospective analysis of women with node-negative breast cancer randomized to receive 5 years of tamoxifen (North Central Cancer Treatment Group 89-30-52). CYP2D6 metabolism (extensive or decreased) was based on CYP2D6*4 genotype and presence/absence of a CYP2D6 inhibitor. Reverse transcription-PCR profiles for HOXB13 and IL17BR and the cut point separating patients into high- and low-risk categories according to disease-free survival (DFS) were used. A risk factor (CYP2D6:HOXB13/IL17BR) representing the four categories of combining CYP2D6 metabolism (extensive or decreased) and HOXB13/IL17BR (low or high) was created. The association between CYP2D6:HOXB13/IL17BR and DFS and overall survival (OS) was assessed using the log-rank test and proportional hazards modeling. CYP2D6 metabolism and HOXB13/IL17BR gene ratio was available in 110 of 160 (69%) patients. The combined CYP2D6:HOXB13/IL17BR risk factor was significantly associated with DFS (log-rank P = 0.004) and OS (P = 0.009). Relative to women with extensive CYP2D6 metabolism and low HOXB13/IL17BR, those with either decreased metabolism or a high HOXB13/IL17BR ratio had significantly worse OS (adjusted hazard ratio, 2.41; 95% confidence interval, 1.08-5.37; P = 0.031), whereas women with both decreased metabolism and high HOXB13/IL17BR ratio had the shortest survival (adjusted hazard ratio, 3.15; 95% CI, 1.17-8.52; P = 0.024). An index composed of inherited (CYP2D6) and tumor (HOXB13/IL17BR) gene variation identifies patients with varying degrees of resistance to tamoxifen.
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.