  • Search Results
  • Impact of obesity control on circulating level of endothelial progenitor cells and angiogenesis in response to ischemic stimulation.
Impact of obesity control on circulating level of endothelial progenitor cells and angiogenesis in response to ischemic stimulation.
Journal of translational medicine (2012-05-10)
Yung-Lung Chen, Chia-Lo Chang, Cheuk-Kwan Sun, Chiung-Jen Wu, Tzu-Hsien Tsai, Sheng-Ying Chung, Sarah Chua, Kuo-Ho Yeh, Steve Leu, Jiunn-Jye Sheu, Fan-Yen Lee, Chia-Hung Yen, Hon-Kan Yip
ABSTRACT
We tested the hypothesis that obesity reduced circulating number of endothelial progenitor cells (EPCs), angiogenic ability, and blood flow in ischemic tissue that could be reversed after obesity control. 8-week-old C57BL/6J mice (n=27) were equally divided into group 1 (fed with 22-week control diet), group 2 (22-week high fat diet), and group 3 (14-week high fat diet, followed by 8-week control diet). Critical limb ischemia (CLI) was induced at week 20 in groups 2 and 3. The animals were sacrificed at the end of 22 weeks. Heart weight, body weight, abdominal fat weight, serum total cholesterol level, and fasting blood sugar were highest in group 2 (all p<0.001). The numbers of circulating EPCs (C-kit/CD31+, Sca-1/KDR + and CXCR4/CD34+) were lower in groups 1 and 2 than in group 3 at 18 h after CLI induction (p<0.03). The numbers of differentiated EPCs (C-kit/CD31+, CXCR4/CD34+ and CD133+) from adipose tissue after 14-day cultivation were also lowest in group 2 (p<0.001). Protein expressions of VCAM-1, oxidative index, Smad3, and TGF-β were higher, whereas the Smad1/5 and BMP-2, mitochondrial cytochrome-C SDF-1α and CXCR4 were lower in group 2 than in groups 1 and 3 (all p<0.02). Immunofluorescent staining of CD31+ and vWF + cells, the number of small vessel (<15 μm), and blood flow through Laser Doppler scanning of ischemic area were lower in group 2 compared to groups 1 and 3 on day 14 after CLI induction (all p<0.001). Obesity suppressed abilities of angiogenesis and recovery from CLI that were reversed by obesity control.
Sigma-Aldrich
OxyBlot Protein Oxidation Detection Kit, The OxyBlot Protein Oxidation Detection Kit provides the reagents to perform the immunoblot detection of carbonyl groups introduced into proteins by oxidative reactions with ozone or oxides of nitrogen or by metal catalyzed oxidation.