  • Search Results
  • Sequestration-based bistability enables tuning of the switching boundaries and design of a latch.
Sequestration-based bistability enables tuning of the switching boundaries and design of a latch.
Molecular systems biology (2012-10-24)
David Chen, Adam P Arkin
ABSTRACT
Natural biological systems have evolved a diverse array of switches to realize their strategies for environmental response and development. Emerging applications of synthetic biology have begun to exploit such switches to achieve increasingly sophisticated designed behaviors. However, not all switch architectures allow facile design of the switching and memory properties. Furthermore, not all designs are built from components for which large families of variants exist, a requirement for building many orthogonal switch variants. Therefore, there is a critical need from genetic engineers for scalable strategies that yield custom bistable switches. Here, we use a sigma factor and its cognate anti-sigma factor to experimentally verify that ultrasensitivity from sequestration combined with positive feedback is sufficient to build a bistable switch. We show that sequestration allows us to predictably tune the switching boundaries, and we can easily tune our switch to function as a set-reset latch that can be toggled between two states by a pulse of inducer input.
Sigma-Aldrich
M9, Minimal Salts, 5X, powder, minimal microbial growth medium
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.