  • Search Results
  • Disabled homolog 2 is required for migration and invasion of prostate cancer cells.
Disabled homolog 2 is required for migration and invasion of prostate cancer cells.
Frontiers of medicine (2015-07-06)
Yinyin Xie, Yuanliang Zhang, Lu Jiang, Mengmeng Zhang, Zhiwei Chen, Dan Liu, Qiuhua Huang
ABSTRACT
Disabled homolog 2 (DAB2) is frequently deleted or epigenetically silenced in many human cancer cells. Therefore, DAB2 has always been regarded as a tumor suppressor gene. However, the role of DAB2 in tumor progression and metastasis remains unclear. In this study, DAB2 expression was upregulated along with human prostate cancer (PCa) progression. DAB2 overexpression or knockdown effects in LNCaP and PC3 cell lines were verified to address the biological functions of DAB2 in PCa progression and metastasis. LNCaP and PC3 cell lines were generated from human PCa cells with low and high metastatic potentials, respectively. The results showed that DAB2 shRNA knockdown can inhibit the migratory and invasive abilities of PC3 cells, as well as the tumorigenicity, whereas DAB2 overexpression enhanced LNCaP cell migration and invasion. Further investigation showed that DAB2 regulated the cell migration associated genes in PC3 cells, and the differential DAB2 expression between LNCaP and PC3 cells was partly regulated by histone 4 acetylation. Therefore, DAB2 may play an important role in PCa progression and metastasis.
Sigma-Aldrich
C646, ≥98% (HPLC)