  • Search Results
  • Ionic liquid polymer functionalized carbon nanotubes-doped poly(3,4-ethylenedioxythiophene) for highly-efficient solid-phase microextraction of carbamate pesticides.
Ionic liquid polymer functionalized carbon nanotubes-doped poly(3,4-ethylenedioxythiophene) for highly-efficient solid-phase microextraction of carbamate pesticides.
Journal of chromatography. A (2016-04-03)
Mian Wu, Liying Wang, Baizhao Zeng, Faqiong Zhao
ABSTRACT
A poly(3,4-ethylenedioxythiophene)-ionic liquid polymer functionalized multiwalled carbon nanotubes (PEDOT-PIL/MWCNTs) composite solid-phase microextraction (SPME) coating was fabricated by electrodeposition. After being dipped in Nafion solution, a Nafion-modified coating was obtained. The outer layer Nafion played a crucial role in enhancing the durability and stability of the coating, thus it was robust enough for replicated extraction for at least 150 times without decrease of extraction performance. The Nafion-modified coating exhibited much higher sensitivity than commercial coatings for the direct extraction of carbamate pesticides in aqueous solutions, due to its strong hydrophobic effect and π-π affinity based enrichment. When it was used for the determination of carbamate pesticides in combination with gas chromatography-flame ionization detection, good linearity (correlation coefficients higher than 0.9981), low limits of detection (15.2-27.2 ng/L) and satisfactory precision (relative standard deviation <8.2%, n=5) were achieved. The developed method was applied to the analysis of four carbamate pesticides in apple and lettuce samples, and acceptable recoveries (i.e. 87.5-106.5%) were obtained for the standard addition.
Product Description
Supelco
SPME Portable Field Sampler, coating PDMS
Supelco
SPME/HPLC Interface, rheodyne valve version
Supelco
SPME Sampling Stand, for use with 15 mL vials
Supelco
SPME Portable Field Sampler, coating CAR/PDMS
Supelco
SPME Sampling Stand, for use with 4 mL vials
Supelco
SPME Portable Field Sampler, coating PDMS/DVB
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm (CAR/PDMS), for use with manual holder, needle size 24 ga
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 24 ga, for use with manual holder
Supelco
SPME fiber assembly polyacrylate (PA), df 85 μm (PA), for use with manual holder, needle size 24 ga
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm (PDMS), needle size 24 ga, for use with manual holder
Supelco
SPME fiber assembly, Carbowax-Polyethylene Glycol (PEG) Coating, needle size 23 ga, df 60 μm (PEG), metal alloy fiber, for use with manual holder
Supelco
SPME fiber assortment kit 1, for use with manual holder, needle size 24 ga
Supelco
SPME fiber assortment kit 2, for use with manual holder, needle size 24 ga
Supelco
SPME fiber assortment kit 4, for use with manual holder, size 24 ga
Supelco
SPME fiber assortment kit 5, needle size 23 ga, for use with autosampler
Supelco
SPME LC Tips, C18, tray of 96 ea
Supelco
SPME StableFlex fiber assortment kit, for use with manual holder
Supelco
SPME-OC Fiber Assembly, Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 75 μm (Coating thickness includes 65 μm coating + 10 μm OC (overcoating)), needle size 23 ga, pkg of 3 ea, fiber L 1 cm
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm (CAR/PDMS), needle size 23 ga, for use with manual holder
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 24 ga, StableFlex, for use with manual holder/autosampler
Supelco
SPME fiber assembly polyacrylate (PA), df 85 μm (PA), for use with autosampler, needle size 24 ga
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm (PDMS), needle size 24 ga, for use with autosampler
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm (PDMS/DVB), needle size 24 ga, for use with manual holder
Supelco
SPME fiber assembly, Carbowax-Polyethylene Glycol (PEG) Coating, needle size 23 ga, df 60 μm (PEG), for use with autosampler
Supelco
SPME fiber assortment kit 1, for use with autosampler, needle size 24 ga
Supelco
SPME fiber assortment kit 2, for use with autosampler, size 24 ga
Supelco
SPME fiber assortment kit 4, for use with autosampler, size 24 ga
Supelco
SPME LC Tips, PDMS/DVB, pkg of 96 ea
Supelco
SPME StableFlex fiber assortment kit, needle size 24 ga, for use with autosampler
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm (CAR/PDMS), needle size 24 ga, for use with autosampler