  • Search Results
  • Early effects of low benzene exposure on blood cell counts in Bulgarian petrochemical workers.
Early effects of low benzene exposure on blood cell counts in Bulgarian petrochemical workers.
La Medicina del lavoro (2009-04-23)
Angela Cecilia Pesatori, S Garte, T Popov, Tzveta Georgieva, T Panev, M Bonzini, D Consonni, M Carugno, B D Goldstein, Emanuela Taioli, V Fontana, Elena Stagi, P A Bertazzi, D F Merlo
ABSTRACT
Only few studies have examined early hematological effects in human populations exposed to low benzene levels and their findings are controversial. We evaluated hematological outcomes (WBC, neutrophils, lymphocytes, monocytes, eosinophils, basophils, RBC, Hb, HCT MCV, platelets and MPV) in a population of 153 Bulgarian petrochemical workers exposed to benzene (range 0.01-23.9 ppm) and 50 unexposed subjects. Written informed consent was obtained and a self-administered questionnaire used to collect information on current smoking habits, lifestyle, and occupational activities. Exposure assessment was based on personal monitoring sampling the day before phlebotomy. Urinary trans-trans-muconic acid (t,t-MA) was determined at the beginning and end of the work shift. Based on individual airborne benzene measurements, study subjects were categorized in three exposure categories (referents, <1 and > or =1 ppm). Mean values of each hematologic outcomes in each exposure category were compared with the referent group using a multiple linear regression model adjusted for age, gender, current smoking habits and environmental toluene level. The influence of the CYP2E1 (RsaI and DraI) and NQO1 609C>T genetic polymorphisms on differential hematological parameters was also investigated. No dose-response effect was observed for most of the examined hematological outcomes (WBC, lymphocytes, neutrophils, monocytes, RBC, Hb, HCT, MCV, platelets and MPV). The eosinophil count was inversely related to benzene exposure only among smokers. Conversely, basophils increased with increasing exposure. No effect on benzene hematotoxicity was found for any of the investigated polymorphisms. In our study we did not find a decline in WBC and lymphocytes related to benzene exposure. A myeloproliferative effect of benzene is highly unlikely to explain the observed reduction in eosinophils and increase in basophils as it would lead to a concordant depression in all granulocyte subpopulations. Whether benzene effects at low doses are present in Caucasian populations remains uncertain, thus warranting further investigations.
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.