  • Search Results
  • Novel organic?inorganic hybrid material based on tris(2,2?-bipyridyl)dichlororuthenium(II) hexahydrate and Dawson-type tungstophosphate K7[H4PW18O62]?18H2O as a bifuctional hydrogen peroxide electrocatalyst for biosensors
Novel organic?inorganic hybrid material based on tris(2,2?-bipyridyl)dichlororuthenium(II) hexahydrate and Dawson-type tungstophosphate K7[H4PW18O62]?18H2O as a bifuctional hydrogen peroxide electrocatalyst for biosensors
Sensors and Actuators B, Chemical
Ammam M and Easton EB