  • Search Results
  • SIRT7 is a histone desuccinylase that functionally links to chromatin compaction and genome stability.
SIRT7 is a histone desuccinylase that functionally links to chromatin compaction and genome stability.
Nature communications (2016-07-21)
Lei Li, Lan Shi, Shangda Yang, Ruorong Yan, Di Zhang, Jianguo Yang, Lin He, Wanjin Li, Xia Yi, Luyang Sun, Jing Liang, Zhongyi Cheng, Lei Shi, Yongfeng Shang, Wenhua Yu
ABSTRACT
Although SIRT7 is a member of sirtuin family proteins that are described as NAD(+)-dependent class III histone deacetylases, the intrinsic enzymatic activity of this sirtuin protein remains to be investigated and the cellular function of SIRT7 remains to be explored. Here we report that SIRT7 is an NAD(+)-dependent histone desuccinylase. We show that SIRT7 is recruited to DNA double-strand breaks (DSBs) in a PARP1-dependent manner and catalyses desuccinylation of H3K122 therein, thereby promoting chromatin condensation and DSB repair. We demonstrate that depletion of SIRT7 impairs chromatin compaction during DNA-damage response and sensitizes cells to genotoxic stresses. Our study indicates SIRT7 is a histone desuccinylase, providing a molecular basis for the understanding of epigenetic regulation by this sirtuin protein. Our experiments reveal that SIRT7-catalysed H3K122 desuccinylation is critically implemented in DNA-damage response and cell survival, providing a mechanistic insight into the cellular function of SIRT7.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone B-5-1-2, purified immunoglobulin, buffered aqueous solution