  • Search Results
  • The metastasis gene NEDD9 product acts through integrin β3 and Src to promote mesenchymal motility and inhibit amoeboid motility.
The metastasis gene NEDD9 product acts through integrin β3 and Src to promote mesenchymal motility and inhibit amoeboid motility.
Journal of cell science (2012-02-14)
Jessica Ahn, Victoria Sanz-Moreno, Christopher J Marshall
ABSTRACT
Neural precursor expressed, developmentally down-regulated 9 (NEDD9), a member of the Cas family of signal transduction molecules, is amplified at the genetic level in melanoma, and elevated expression levels have been shown to correlate with melanoma progression and metastasis. NEDD9 interacts with the guanine nucleotide exchange factor DOCK3 to promote Rac activation and the elongated, mesenchymal-type of tumour cell invasion, but the molecular mechanisms through which NEDD9 promotes melanoma metastasis are not fully understood. We show that signalling through increased NEDD9 levels requires integrin β3 signalling, which leads to elevated phosphorylation of integrin β3. This results in increased Src and FAK but decreased ROCK signalling to drive elongated, mesenchymal-type invasion in environments that contain vitronectin. NEDD9 overexpression does not affect ROCK signalling through activation of RhoA but decreases ROCKII signalling through Src-dependent phosphorylation of a negative regulatory site Tyr722. In NEDD9-overexpressing melanoma cells, inhibition of Src with dasatinib results in a switch from Rac-driven elongated, mesenchymal-type invasion to ROCK-dependent rounded, amoeboid invasion. These findings brings into question whether dasatinib would work as a therapeutic agent to block melanoma invasion and metastasis. On the basis of the in vitro data presented here, a combination treatment of dasatinib and a ROCK inhibitor might be a better alternative in order to inhibit both elongated, mesenchymal-type and rounded, amoeboid motility.
Sigma-Aldrich
Monoclonal Anti-Myosin (Light Chains 20 kDa) antibody produced in mouse, clone MY-21, ascites fluid
Sigma-Aldrich
Anti-Vitronectin Antibody, clone BV2, clone BV2, Chemicon®, from mouse
Sigma-Aldrich
4G10® Platinum, Anti-Phosphotyrosine Antibody (mouse monoclonal cocktail IgG2b), clone 4G10®, Upstate®, from mouse
Sigma-Aldrich
Anti-Rac1 Antibody, clone 23A8, clone 23A8, Upstate®, from mouse
Sigma-Aldrich
Anti-Integrin αVβ3 Antibody, clone LM609, Azide Free, clone LM609, Chemicon®, from mouse