  • Search Results
  • Enhanced autointegration in hyperstable simian immunodeficiency virus capsid mutants blocked after reverse transcription.
Enhanced autointegration in hyperstable simian immunodeficiency virus capsid mutants blocked after reverse transcription.
Journal of virology (2013-01-25)
Christopher Tipper, Joseph Sodroski
ABSTRACT
After entering a host cell, retroviruses such as simian immunodeficiency virus (SIV) uncoat, disassembling the viral capsid. Rates of uncoating that are too high and too low can be detrimental to the efficiency of infection. Rapid uncoating typically leads to blocks in reverse transcription, but the basis for replication defects associated with slow uncoating is less clear. Here we characterize the phenotypes of two SIVmac239 mutants with changes, A87E and A87D, in the helix 4/5 loop of the capsid protein. These mutant viruses exhibited normal capsid morphology but were significantly attenuated for infectivity. The infectivity of wild-type and mutant SIVmac239 was not decreased by aphidicolin-induced growth arrest of the target cells. In the cytosol of infected cells, the A87E and A87D capsids remained in particulate form longer than the wild-type SIVmac239 capsid, suggesting that the mutants uncoat more slowly than the wild-type capsid. Both mutants exhibited much higher levels of autointegrated DNA forms than wild-type SIVmac239. Thus, some changes in the helix 4/5 loop of the SIVmac239 capsid protein result in capsid hyperstability and an increase in autointegration.
Sigma-Aldrich
Anti-Human IgG (Fab specific)−Peroxidase antibody produced in goat, affinity isolated antibody