  • Search Results
  • Non-pheochromocytoma (PCC)/paraganglioma (PGL) tumors in patients with succinate dehydrogenase-related PCC-PGL syndromes: a clinicopathological and molecular analysis.
Non-pheochromocytoma (PCC)/paraganglioma (PGL) tumors in patients with succinate dehydrogenase-related PCC-PGL syndromes: a clinicopathological and molecular analysis.
European journal of endocrinology (2013-10-08)
Thomas G Papathomas, Jose Gaal, Eleonora P M Corssmit, Lindsey Oudijk, Esther Korpershoek, Ketil Heimdal, Jean-Pierre Bayley, Hans Morreau, Marieke van Dooren, Konstantinos Papaspyrou, Thomas Schreiner, Torsten Hansen, Per Arne Andresen, David F Restuccia, Ingrid van Kessel, Geert J L H van Leenders, Johan M Kros, Leendert H J Looijenga, Leo J Hofland, Wolf Mann, Francien H van Nederveen, Ozgur Mete, Sylvia L Asa, Ronald R de Krijger, Winand N M Dinjens
ABSTRACT
Although the succinate dehydrogenase (SDH)-related tumor spectrum has been recently expanded, there are only rare reports of non-pheochromocytoma/paraganglioma tumors in SDHx-mutated patients. Therefore, questions still remain unresolved concerning the aforementioned tumors with regard to their pathogenesis, clinicopathological phenotype, and even causal relatedness to SDHx mutations. Absence of SDHB expression in tumors derived from tissues susceptible to SDH deficiency is not fully elucidated. Three unrelated SDHD patients, two with pituitary adenoma (PA) and one with papillary thyroid carcinoma (PTC), and three SDHB patients affected by renal cell carcinomas (RCCs) were identified from four European centers. SDHA/SDHB immunohistochemistry (IHC), SDHx mutation analysis, and loss of heterozygosity analysis of the involved SDHx gene were performed on all tumors. A cohort of 348 tumors of unknown SDHx mutational status, including renal tumors, PTCs, PAs, neuroblastic tumors, seminomas, and adenomatoid tumors, was investigated by SDHB IHC. Of the six index patients, all RCCs and one PA displayed SDHB immunonegativity in contrast to the other PA and PTC. All immunonegative tumors demonstrated loss of the WT allele, indicating bi-allelic inactivation of the germline mutated gene. Of 348 tumors, one clear cell RCC exhibited partial loss of SDHB expression. These findings strengthen the etiological association of SDHx genes with pituitary neoplasia and provide evidence against a link between PTC and SDHx mutations. Somatic deletions seem to constitute the second hit in SDHB-related renal neoplasia, while SDHx alterations do not appear to be primary drivers in sporadic tumorigenesis from tissues affected by SDH deficiency.
Sigma-Aldrich
Anti-SDHB antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution