  • Search Results
  • Increased KPI containing amyloid precursor protein in experimental autoimmune encephalomyelitis brains.
Increased KPI containing amyloid precursor protein in experimental autoimmune encephalomyelitis brains.
Neuroreport (2007-04-07)
Orit Beilin, Dimitrios M Karussis, Amos D Korczyn, David Gurwitz, Ramona Aronovich, Rachel Mizrachi-Kol, Joab Chapman
ABSTRACT
Amyloid precursor protein can be translated from three alternatively spliced mRNAs. We measured levels of amyloid precursor protein isoforms containing the Kunitz protease inhibitor domain (KPIAPP), and amyloid precursor protein without the Kunitz protease inhibitor domain (KPIAPP) in brain homogenates of acute experimental autoimmune encephalomyelitis mice. At the preclinical phase of the disease, both KPIAPP and KPIAPP levels were significantly higher in homogenates from brains of autoimmune encephalomyelitis mice, whereas at the acute phase of the disease only KPIAPP remained significantly elevated compared with controls. At the recovery phase, no differences were observed between the groups. The early and isoform-specific elevation of KPIAPP in autoimmune encephalomyelitis mice suggests a possible role for amyloid precursor protein in the immune response mediating the disease.
Sigma-Aldrich
Anti-Amyloid Precursor Protein, KPI Domain antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.