  • Search Results
  • The genetics of Canadian type 3 von Willebrand disease: further evidence for co-dominant inheritance of mutant alleles.
The genetics of Canadian type 3 von Willebrand disease: further evidence for co-dominant inheritance of mutant alleles.
Journal of thrombosis and haemostasis : JTH (2013-01-15)
M Bowman, A Tuttle, C Notley, C Brown, S Tinlin, M Deforest, J Leggo, V S Blanchette, D Lillicrap, P James
ABSTRACT
Type 3 von Willebrand disease (VWD) is the most severe form of the disease and is classically inherited in an autosomal recessive fashion. The aim of the current study was to investigate the molecular pathogenesis of a Canadian cohort of type 3 VWD patients. Thirty-four families comprised of 100 individuals were investigated. Phenotypic data, including bleeding scores (BS), von Willebrand factor (VWF) laboratory values and anti-VWF inhibitor status were included as well as sequence analysis. We identified 31 different mutations (20 novel): 8 frameshift, 5 splice site, 9 nonsense, 1 gene conversion, 6 missense and 2 partial gene deletion mutations. The majority of mutations identified were in the propeptide (42%); index cases (IC) with these mutations exhibited more severe bleeding (BS = 22) than those with mutations elsewhere in VWF (BS = 13). Sixty-two out of 68 (91%) mutant alleles were identified. Twenty-nine IC (85%) had a VWF null genotype identified; 17 homozygous, 12 compound heterozygous. In five IC (15%), two mutant VWF alleles were not identified to explain the type 3 VWD phenotype. In four ICs only one mutant VWF allele was identified and in one IC no mutant VWF alleles were identified. We have investigated the molecular pathogenesis of a Canadian cohort of type 3 VWD patients. Obligate carriers are not phenotypically silent in the Canadian population; 48% have been diagnosed with type 1 VWD. In approximately 50% of families in this study the inheritance pattern for type 3 VWD is co-dominant and not recessive.
Sigma-Aldrich
Anti-Human IgG (γ-chain specific), F(ab′)2 fragment−Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.