  • Search Results
  • Integrin alphavbeta6 mediates HT29-D4 cell adhesion to MMP-processed fibrinogen in the presence of Mn2+.
Integrin alphavbeta6 mediates HT29-D4 cell adhesion to MMP-processed fibrinogen in the presence of Mn2+.
European journal of cell biology (2007-02-06)
Francis Fouchier, Claude Penel, Marie Pierre Montero, Patricia Bremond, Serge Champion
ABSTRACT
Mn(2+) was found to induce adhesion of HT29-D4 adenoma carcinoma cells to fibrinogen (Fb). This was independent of the expression of the beta3 integrin subunit and involved endogenous alphavbeta6 but not alphavbeta5 integrin. Thus, addition of Mn(2+) led to a change in integrin alphavbeta6 specificity. Furthermore, Mn(2+) was found to strongly activate the extracellular signal-regulated kinase/mitogen-activated protein kinase (ERK/MAPK) pathway in the HT29-D4 cell line. As a MAPK inhibitor strongly reduced the Mn(2+)-induced cell adhesion to Fb, it is suggested that a link between MAPK activation and cell adhesion to Fb exists. Both expression and activity of matrix metalloproteinase-9 (MMP-9) were enhanced by Mn(2+) and this led to Fb processing. MMP inhibitors prevented Mn(2+)-mediated cell adhesion to Fb, leading us to suggest that Mn(2+) promoted convergent changes in integrin alphavbeta6 conformation and Fb structure through activation of ERK/MAPK and MMP-9. Finally, we found that Mn(2+) and activators of the ERK pathway cooperated in HT29-D4 cell adhesion to Fb. Such a process may be involved in bone metastasis of some cancer cells.
Sigma-Aldrich
Anti-Fibrinogen antibody produced in goat, whole antiserum
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.