  • Search Results
  • Localization of beta-secretase memapsin 2 in the brain of Alzheimer's patients and normal aged controls.
Localization of beta-secretase memapsin 2 in the brain of Alzheimer's patients and normal aged controls.
Experimental neurology (2002-05-16)
Anyang Sun, Gerald Koelsch, Jordan Tang, Guoying Bing
ABSTRACT
Chronic accumulation of beta-amyloid in the brain has been shown to result in complex molecular and cellular changes that accompany neurodegeneration in Alzheimer's disease (AD). In this study, we examined the expression of a newly identified beta-secretase, memapsin 2 (M2) or beta-site APP cleaving enzyme in deparaffinized sections from 10 AD patients and 10 aged matched controls and in frozen samples of parietal cortex from 11 AD and 8 controls. M2 is mainly expressed in neurons, with high levels in CA4 to CA2 regions and transentorhinal cortex and low or intermediate levels in CA1, subiculum, and granule cells of the dentate gyrus. The majority of AD brains showed an increase of M2 expression in the CA1, but a decrease in the transentorhinal cortex. A subset of controls and AD patients had high M2 expression in parietal neocortex. Double-staining revealed that senile plaques are not directly associated with the soma of M2-expressing neurons. Neurofibrillary tangles were associated with lower M2 expression in AD. These data indicate that beta-secretase M2 may not be straightforwardly involved in amyloid plaque formation in AD brain.
Sigma-Aldrich
Monoclonal Anti-β-Amyloid antibody produced in mouse, clone BAM-10, ascites fluid
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.