  • Search Results
  • Effect of immunoglobulin light chains from hemodialysis and continuous ambulatory peritoneal dialysis patients on polymorphonuclear leukocyte functions.
Effect of immunoglobulin light chains from hemodialysis and continuous ambulatory peritoneal dialysis patients on polymorphonuclear leukocyte functions.
Journal of the American Society of Nephrology : JASN (1995-12-01)
G Cohen, M Haag-Weber, B Mai, R Deicher, W H Hörl
ABSTRACT
Circulating plasma factors accumulating in the serum of uremic patients have the potential to inhibit essential functions of polymorphonuclear leukocytes (PMNL). As a consequence, these factors can contribute to the increased risk for bacterial infections generally found in uremic patients. Free immunoglobulin light chains that are present in the serum of healthy adults at low levels appear in the serum of uremic patients at significantly higher levels. Therefore, kappa and lambda light chains in their monomeric and dimeric forms were isolated from hemodialysis and continuous ambulatory peritoneal dialysis patients and their potential to inhibit essential PMNL functions in in vitro assays was tested. It was found that all isolates tested were able to inhibit deoxyglucose uptake, a measure for the state of activation of PMNL, as well as chemotaxis. In contrast, free immunoglobulin light chains had no influence on the phagocytotic functions of PMNL. It was concluded that free immunoglobulin light chains are able to act as uremic toxins by interfering with essential PMNL functions and that their serum levels and fate during the treatment of uremic patients should be taken into consideration.
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.