  • Search Results
  • Antinucleosome antibodies in primary antiphospholipid syndrome: a hint at systemic autoimmunity?
Antinucleosome antibodies in primary antiphospholipid syndrome: a hint at systemic autoimmunity?
Journal of autoimmunity (2008-01-15)
Laura Andreoli, Francesca Pregnolato, Rufus W Burlingame, Flavio Allegri, Silvia Rizzini, Valentina Fanelli, Antonella Radice, Caterina Corace, Renato Alberto Sinico, Pier Luigi Meroni, Angela Tincani
ABSTRACT
Antinucleosome antibodies (anti-NCS) are reported to be highly sensitive and specific for systemic lupus erythematosus (SLE) and to correlate with disease activity. They may appear in early stages of the disease, in particular before anti-dsDNA antibodies, being a potential marker for identifying patients susceptible to SLE. Patients with primary antiphospholipid syndrome (PAPS) may develop full-blown SLE but there is no evidence for markers predictive for that. To evaluate whether anti-NCS may be predictors for full-blown or lupus like disease (LL) in a cohort of PAPS patients. A multicentric cohort of 105 PAPS patients was tested for IgG/IgM anti-NCS by using a home made assay with H1-stripped chromatin as antigen. Eighty-one out of 105 (77%) of the patients were positive for anti-NCS; medium-high titre results were present only in 49/105 (46%). Anti-NCS were more frequently detected in PAPS+LL, but no relationship with clinical/serological features was found, except for a weak correlation with anti-dsDNA antibodies. Two PAPS patients evolved into full-blown SLE during the follow-up and displayed high titre anti-NCS many years before. Our findings suggest that anti-NCS might be added to the mosaic of autoimmune phenomena characterizing PAPS patients and in particular those with more chance to evolve to SLE.
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.