  • Search Results
  • CREB regulates AChE-R-induced proliferation of human glioblastoma cells.
CREB regulates AChE-R-induced proliferation of human glioblastoma cells.
Neoplasia (New York, N.Y.) (2004-05-22)
Chava Perry, Ella H Sklan, Hermona Soreq
ABSTRACT
The cyclic adenosine monophosphate (AMP) response element-binding protein, CREB, often modulates stress responses. Here, we report that CREB suppresses the glioblastoma proliferative effect of the stress-induced acetylcholinesterase variant, AChE-R. In human U87MG glioblastoma cells, AChE-R formed a triple complex with protein kinase C (PKC) epsilon and the scaffold protein RACK1, enhanced PKCepsilon phosphorylation, and facilitated BrdU incorporation. Either overexpressed CREB, or antisense destruction of AChE-R mRNA, PKC, or protein kinase A (PKA) inhibitors-but not CREB combined with PKC inhibition suppressed-this proliferation, suggesting that CREB's repression of this process involves a PKC-mediated pathway, whereas impaired CREB regulation allows AChE-R-induced, PKA-mediated proliferation of glioblastoma tumors.
Sigma-Aldrich
Monoclonal Anti-Protein Kinase Cβ2 antibody produced in mouse, clone PK-B26, ascites fluid
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.