  • Search Results
  • Ligand-specific function of transforming growth factor beta in epithelial-mesenchymal transition in heart development.
Ligand-specific function of transforming growth factor beta in epithelial-mesenchymal transition in heart development.
Developmental dynamics : an official publication of the American Association of Anatomists (2009-01-24)
Mohamad Azhar, Raymond B Runyan, Connie Gard, L Philip Sanford, Marian L Miller, Anastasia Andringa, Sharon Pawlowski, Sudarsan Rajan, Thomas Doetschman
ABSTRACT
The ligand specificity of transforming growth factor beta (TGFbeta) in vivo in mouse cardiac cushion epithelial-to-mesenchymal transition (EMT) is poorly understood. To elucidate the function of TGFbeta in cushion EMT, we analyzed Tgfb1(-/-), Tgfb2(-/-), and Tgfb3(-/-) mice between embryonic day (E) 9.5 and E14.5 using both in vitro and in vivo approaches. Atrioventricular (AV) canal collagen gel assays at E9.5 indicated normal EMT in both Tgfb1(-/-) and Tgfb3(-/-) mice. However, analysis of Tgfb2(-/-) AV explants at E9.5 and E10.5 indicated that EMT, but not cushion cell proliferation, was initially delayed but later remained persistent. This was concordant with the observation that Tgfb2(-/-) embryos, and not Tgfb1(-/-) or Tgfb3(-/-) embryos, develop enlarged cushions at E14.5 with elevated levels of well-validated indicators of EMT. Collectively, these data indicate that TGFbeta2, and not TGFbeta1 or TGFbeta3, mediates cardiac cushion EMT by promoting both the initiation and cessation of EMT.
Sigma-Aldrich
Ras Activation Assay Kit
Sigma-Aldrich
Monoclonal Anti-MAP Kinase, Activated (Diphosphorylated ERK-1&2) antibody produced in mouse, clone MAPK-YT, ascites fluid
Sigma-Aldrich
Anti-Fibronectin Antibody, Chemicon®, from rabbit