MilliporeSigma
  • Drosophila UTX is a histone H3 Lys27 demethylase that colocalizes with the elongating form of RNA polymerase II.
Drosophila UTX is a histone H3 Lys27 demethylase that colocalizes with the elongating form of RNA polymerase II.
Molecular and cellular biology (2007-11-28)
Edwin R Smith, Min Gyu Lee, Benjamin Winter, Nathan M Droz, Joel C Eissenberg, Ramin Shiekhattar, Ali Shilatifard
ABSTRACT
Histone H3 methylation at Lys27 (H3K27 methylation) is a hallmark of silent chromatin, while H3K4 methylation is associated with active chromatin regions. Here we report that a Drosophila JmjC family member, dUTX, specifically demethylates di- and trimethylated but not monomethylated H3K27. dUTX localization on chromatin correlates with the elongating form of RNA polymerase II (Pol II), and dUTX can associate with Pol II. Furthermore, heat shock induction results in the recruitment of dUTX to the hsp70 gene, like that of several other Pol II elongation factors. Our data indicate that dUTX is intimately associated with actively transcribed genes and may provide a paradigm for how H3K27 demethylation is required for the activation of preinitiated Pol II on transcriptionally poised genes.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys4) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-monomethyl-Histone H3 (Lys27) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys9) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-dimethyl-Histone H3 (Lys27) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys27) Antibody, Upstate®, from rabbit