  • Search Results
  • A discrepancy between liquid phase and gel phase assays for evaluation of total complement activity and some possible explanations.
A discrepancy between liquid phase and gel phase assays for evaluation of total complement activity and some possible explanations.
Scandinavian journal of immunology (2004-06-09)
S Thordardottir, K H Traustadottir, K Erlendsson
ABSTRACT
This study was conducted to investigate the frequency and origin of discrepant assay results between two haemolytic assays which both measure activity of the classical pathway of complement (CH50) by haemolysis of sheep red blood cells (SRBCs). One is conducted in gel phase using undiluted sera and the other in liquid phase with sera in 1/100 dilution. The majority of discrepant readings are observed as low or absent haemolysis in the gel phase, with values within or above the normal range in the liquid phase. The incidence of discrepant assay readings was evaluated in 300 samples. Furthermore, 28 samples showing the most discrepant readings were investigated further for disturbing factors. Factors evaluated in the test sera were mannose-binding lectin, C-reactive protein (CRP), immune complexes, antibodies to SRBCs, rheumatoid factor and immunoglobulin A (IgA) and IgG anti-C1q antibodies. The results showed that discrepant readings are present in 10% of the 300 samples and false low gel assay readings account for 6.3%. The majority (68%) of the discrepant samples contained a heat-stable-inhibiting factor, and the main mediators found were elevated levels of IgA anti-C1q antibodies and antibodies to SRBCs. This could indicate a clinically relevant factor in the test sera but can also result from the difference in assay design.
Sigma-Aldrich
Anti-Human IgG−Alkaline Phosphatase antibody, Mouse monoclonal, clone GG-5, purified from hybridoma cell culture
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.