  • Search Results
  • A phase II study of enzastaurin, a protein kinase C beta inhibitor, in patients with relapsed or refractory mantle cell lymphoma.
A phase II study of enzastaurin, a protein kinase C beta inhibitor, in patients with relapsed or refractory mantle cell lymphoma.
Annals of oncology : official journal of the European Society for Medical Oncology (2007-10-02)
F Morschhauser, J F Seymour, H C Kluin-Nelemans, A Grigg, M Wolf, M Pfreundschuh, H Tilly, J Raemaekers, M B van 't Veer, N Milpied, G Cartron, A Pezzutto, A Spencer, F Reyes, M Dreyling
ABSTRACT
Protein kinase C beta (PKCbeta), a pivotal enzyme in B-cell signaling and survival, is overexpressed in most cases of mantle cell lymphoma (MCL). Activation of PI3K/AKT pathway is involved in pathogenesis of MCL. Enzastaurin, an oral serine/threonine kinase inhibitor, suppresses signaling through PKCbeta/PI3K/AKT pathways, induces apoptosis, reduces proliferation, and suppresses tumor-induced angiogenesis. Patients with relapsed/refractory MCL, and no more than four regimens of prior therapy, received 500 mg enzastaurin, orally, once daily. Sixty patients, median age 66 years (range 45-85), Eastern Cooperative Oncology Group performance status of zero to two (48% had baseline International Prognostic Index of 3-5), were enrolled. Most patients had prior CHOP-like chemotherapy and/or rituximab (median = 2 regimens). No drug-related deaths occurred. There was one case each of grade 3 anemia, diarrhea, dyspnea, vomiting, hypotension, and syncope. Fatigue was the most common toxicity. Although no objective tumor responses occurred, 22 patients (37%, 95% CI 25% to 49%) were free from progression (FFP) for > or =3 cycles (one cycle = 28 days); 6 of 22 were FFP for >6 months. Two patients remain on treatment and FFP at >23 months. Freedom from progression for >6 months in six patients and a favorable toxicity profile with minimal hematological toxicity indicate that enzastaurin warrants evaluation as maintenance therapy and combination chemotherapy in MCL.
Sigma-Aldrich
Monoclonal Anti-Protein Kinase Cβ2 antibody produced in mouse, clone PK-B26, ascites fluid
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.