An algorithm for family screening for coeliac disease.
World journal of gastroenterology (2007-01-05)
Jocelyn-S Fraser, Alistair-L King, H-Julia Ellis, Simon-J Moodie, Ingvar Bjarnason, Jill Swift, Paul-J Ciclitira
ABSTRACT
To assess the level of undiagnosed coeliac disease (CD) in relatives of patients affected by the condition. We collected blood from 914 relatives of probands. We screened these individuals by ELISA for IgA and IgG tTG antibodies, confirming any positive IgA tTG results with an IgA EMA and looked for evidence of IgA deficiency in those who were IgG tTG positive alone, and performed IgG1 EMA in these individuals. We undertook HLA typing where positive screening was found, and this confirmed a strong prevalence of HLA-DQ2 in the coeliac population. Follow-up small intestinal biopsy was undertaken in cases with positive serological screening, wherever possible. Use of this serological screening algorithm revealed a prevalence of undiagnosed CD in 5%-6% of first degree relatives of probands. Our data suggests that first degree relatives of individuals with CD should be screened for this condition.
Sigma-Aldrich
Monoclonal Anti-Human IgG1−FITC antibody produced in mouse, clone 8c/6-39, purified immunoglobulin, buffered aqueous solution
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.