US
EN
Applications
Products
Services
Support
Home
Search Results
A thermodynamic model for Nap1-histone interactions.
A thermodynamic model for Nap1-histone interactions.
The Journal of Biological Chemistry (2008)
Andrews AJ, et al.