US
EN
Applications
Products
Services
Support
Home
Search Results
Matrix-assisted laser desorption ionization mass spectrometry with 2-(4-hydroxyphenylazo) benzoic acid matrix.
Matrix-assisted laser desorption ionization mass spectrometry with 2-(4-hydroxyphenylazo) benzoic acid matrix.
Journal of the American Society For Mass Spectrometry (1993)
Juhasz, Peter, Catherine E. Costello, and Klaus Biemann.